Assay Details
| Gene Symbol: HAF01 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103009.4 | NP_174552.1 | 17 - 18 | 4147 | 72 | |
| GenBank mRNA | AF510669.1 | - | 16 - 17 | 3855 | 72 | |
More Information
Set Membership: |
|

HAF01
Genomic Map