Assay Details
| Gene Symbol: AT1G32930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103026.4 | NP_174569.1 | 5 - 6 | 900 | 140 | |
| GenBank mRNA | AK227878.1 | - | 5 - 6 | 776 | 140 | |
| BT006435.1 | - | 5 - 6 | 540 | 140 | ||
| KJ138975.1 | - | 5 - 6 | 540 | 140 | ||
More Information
Set Membership: |
|

AT1G32930
Genomic Map