Assay Details
| Gene Symbol: AT1G32930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103026.4 | NP_174569.1 | 8 - 9 | 1178 | 103 | |
| GenBank mRNA | AK227878.1 | - | 8 - 9 | 1054 | 103 | |
| BT006435.1 | - | 8 - 9 | 818 | 103 | ||
| KJ138975.1 | - | 8 - 9 | 818 | 103 | ||
More Information
Set Membership: |
|

AT1G32930
Genomic Map