Assay Details
| Gene Symbol: SAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198200.2 | NP_001185129.1 | 12 - 13 | 2388 | 94 | |
| NM_001333042.1 | NP_001319133.1 | 12 - 13 | 2376 | 94 | ||
More Information
Set Membership: |
|

SAR1
Genomic Map