Assay Details
| Gene Symbol: PUM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333166.1 | NP_001322303.1 | 2 - 3 | 1318 | 77 | |
| NM_103275.1 | NP_174811.1 | 2 - 3 | 1183 | 77 | ||
| GenBank mRNA | DQ446324.1 | - | 2 - 3 | 1183 | 77 | |
More Information
Set Membership: |
|

PUM9
Genomic Map