Assay Details
| Gene Symbol: BGAL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333243.1 | NP_001320473.1 | 12 - 13 | 1574 | 90 | |
| NM_103587.3 | NP_175127.1 | 14 - 15 | 2106 | 90 | ||
| GenBank mRNA | AY069911.1 | - | 14 - 15 | 1905 | 90 | |
| AY093977.1 | - | 14 - 15 | 1770 | 90 | ||
More Information
Set Membership: |
|

BGAL5
Genomic Map