Assay Details
| Gene Symbol: BGAL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333243.1 | NP_001320473.1 | 5 - 6 | 584 | 83 | |
| NM_103587.3 | NP_175127.1 | 7 - 8 | 1116 | 83 | ||
| GenBank mRNA | AY069911.1 | - | 7 - 8 | 915 | 83 | |
| AY093977.1 | - | 7 - 8 | 780 | 83 | ||
More Information
Set Membership: |
|

BGAL5
Genomic Map