Assay Details
| Gene Symbol: CIPK17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333316.1 | NP_001320279.1 | 1 - 2 | 339 | 108 | |
| NM_001333318.1 | NP_001320277.1 | 1 - 2 | 374 | 108 | ||
| NM_001333319.1 | NP_001320278.1 | 1 - 2 | 374 | 108 | ||
| NM_103723.4 | NP_175260.1 | 1 - 2 | 339 | 108 | ||
| GenBank mRNA | BT029240.1 | - | 1 - 2 | 159 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001333316.1, NM_001333318.1, NM_001333319.1 transcripts.
Set Membership: |
|

CIPK17
Genomic Map