Assay Details
| Gene Symbol: CIPK17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103723.4 | NP_175260.1 | 2 - 3 | 403 | 144 | |
| GenBank mRNA | AY036958.1 | - | 2 - 3 | 223 | 144 | |
| BT029240.1 | - | 2 - 3 | 223 | 144 | ||
More Information
Set Membership: |
|

CIPK17
Genomic Map