Assay Details
| Gene Symbol: CIPK17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333316.1 | NP_001320279.1 | 4 - 5 | 1554 | 87 | |
| NM_001333317.1 | NP_001320280.1 | 3 - 4 | 785 | 87 | ||
| NM_001333318.1 | NP_001320277.1 | 5 - 6 | 1396 | 87 | ||
| NM_001333319.1 | NP_001320278.1 | 5 - 6 | 1114 | 87 | ||
| NM_103723.4 | NP_175260.1 | 5 - 6 | 668 | 87 | ||
| GenBank mRNA | AY036958.1 | - | 5 - 6 | 488 | 87 | |
| BT029240.1 | - | 5 - 6 | 488 | 87 | ||
More Information
Set Membership: |
|

CIPK17
Genomic Map