Assay Details
| Gene Symbol: BGLU27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115875.5 | NP_191571.4 | 6 - 7 | 664 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BGLU27| Gene Symbol: BGLU27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115875.5 | NP_191571.4 | 6 - 7 | 664 | 95 | |
Set Membership: |
|