Assay Details
| Gene Symbol: CKB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125389.2 | NP_001118861.1 | 3 - 4 | 749 | 104 | |
| NM_001340030.1 | NP_001325549.1 | 3 - 4 | 749 | 104 | ||
| NM_115889.5 | NP_191584.1 | 3 - 4 | 749 | 104 | ||
| GenBank mRNA | AF068318.2 | - | 3 - 4 | 662 | 104 | |
| AK229231.1 | - | 3 - 4 | 617 | 104 | ||
More Information
Set Membership: |
|

CKB3
Genomic Map