Assay Details
| Gene Symbol: AT5G12980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121301.5 | NP_196802.1 | 1 - 2 | 255 | 71 | |
| GenBank mRNA | AY161167.1 | - | 1 - 2 | 199 | 71 | |
More Information
Set Membership: |
|

AT5G12980
Genomic Map