Assay Details
| Gene Symbol: KT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336238.1 | NP_001324058.1 | 2 - 3 | 828 | 136 | |
| NM_128562.4 | NP_180568.1 | 2 - 3 | 523 | 136 | ||
| GenBank mRNA | AF029876.1 | - | 2 - 3 | 294 | 136 | |
| AF033118.1 | - | 2 - 3 | 328 | 136 | ||
| AK221387.1 | - | 2 - 3 | 390 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001336238.1 transcripts.
Set Membership: |
|

KT1
Genomic Map