Assay Details
| Gene Symbol: SAG29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121320.3 | NP_196821.1 | 2 - 3 | 274 | 108 | |
| GenBank mRNA | AY045949.1 | - | 2 - 3 | 227 | 108 | |
| AY113934.1 | - | 2 - 3 | 91 | 108 | ||
More Information
Set Membership: |
|

SAG29
Genomic Map