Assay Details
| Gene Symbol: SAG29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121320.3 | NP_196821.1 | 5 - 6 | 769 | 86 | |
| GenBank mRNA | AY045949.1 | - | 5 - 6 | 722 | 86 | |
| AY113934.1 | - | 5 - 6 | 586 | 86 | ||
More Information
Set Membership: |
|

SAG29
Genomic Map