Assay Details
| Gene Symbol: PBS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001343268.1 | NP_001330075.1 | 3 - 4 | 739 | 99 | |
| NM_001343269.1 | NP_001330076.1 | 2 - 3 | 752 | 99 | ||
| NM_121335.4 | NP_196836.1 | 3 - 4 | 529 | 99 | ||
| GenBank mRNA | EU214910.1 | - | 2 - 3 | 382 | 99 | |
More Information
Set Membership: |
|

PBS3
Genomic Map