Assay Details
| Gene Symbol: ATP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036798.1 | NP_001031875.1 | 1 - 2 | 172 | 62 | |
| NM_121348.4 | NP_196849.1 | 1 - 2 | 226 | 62 | ||
| GenBank mRNA | AK317234.1 | - | 1 - 2 | 172 | 62 | |
More Information
Set Membership: |
|

ATP5
Genomic Map