Assay Details
| Gene Symbol: EMB2773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203388.1 | NP_001190317.1 | 14 - 15 | 2833 | 70 | |
| NM_121558.4 | NP_197058.2 | 14 - 15 | 3094 | 70 | ||
| GenBank mRNA | EF520004.1 | - | 14 - 15 | 2839 | 70 | |
More Information
Set Membership: |
|

EMB2773
Genomic Map