Assay Details
| Gene Symbol: EMB2773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121558.4 | NP_197058.2 | 2 - 3 | 636 | 72 | |
| GenBank mRNA | EF520004.1 | - | 2 - 3 | 381 | 72 | |
More Information
Set Membership: |
|

EMB2773
Genomic Map