Assay Details
| Gene Symbol: RIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121654.3 | NP_197153.1 | 1 - 2 | 142 | 150 | |
| GenBank mRNA | AK176878.1 | - | 1 - 2 | 125 | 150 | |
| BT010523.1 | - | 1 - 2 | 117 | 150 | ||
More Information
Set Membership: |
|

RIC4
Genomic Map