Assay Details
| Gene Symbol: BPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085127.1 | NP_001078596.1 | 2 - 3 | 232 | 85 | |
| NM_121690.4 | NP_197186.1 | 2 - 3 | 317 | 85 | ||
More Information
Set Membership: |
|

BPA1
Genomic Map