Assay Details
| Gene Symbol: MYB56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121786.4 | NP_197282.1 | 2 - 3 | 940 | 68 | |
| GenBank mRNA | AK176193.1 | - | 2 - 3 | 864 | 68 | |
| AK176277.1 | - | 2 - 3 | 864 | 68 | ||
| AY519627.1 | - | 2 - 3 | 806 | 68 | ||
More Information
Set Membership: |
|

MYB56
Genomic Map