Assay Details
| Gene Symbol: TRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121804.5 | NP_197300.1 | 2 - 3 | 586 | 97 | |
| GenBank mRNA | AY054506.1 | - | 2 - 3 | 450 | 97 | |
| AY093298.1 | - | 2 - 3 | 336 | 97 | ||
More Information
Set Membership: |
|

TRP1
Genomic Map