Assay Details
| Gene Symbol: DRT101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121812.3 | NP_568359.2 | 1 | 2116 | 137 | |
| GenBank mRNA | AY075620.1 | - | 1 | 1878 | 137 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DRT101| Gene Symbol: DRT101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121812.3 | NP_568359.2 | 1 | 2116 | 137 | |
| GenBank mRNA | AY075620.1 | - | 1 | 1878 | 137 | |
Set Membership: |
|