Assay Details
| Gene Symbol: APL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121927.3 | NP_197423.1 | 13 - 14 | 1458 | 115 | |
| GenBank mRNA | AF370503.1 | - | 13 - 14 | 1457 | 115 | |
| BT008884.1 | - | 13 - 14 | 1402 | 115 | ||
| U72290.1 | - | 13 - 14 | 1415 | 115 | ||
| X73367.2 | - | 1415 | 115 | |||
More Information
Set Membership: |
|

APL1
Genomic Map