Assay Details
| Gene Symbol: APL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121927.3 | NP_197423.1 | 14 - 15 | 1518 | 121 | |
| GenBank mRNA | AF370503.1 | - | 14 - 15 | 1517 | 121 | |
| BT008884.1 | - | 14 - 15 | 1462 | 121 | ||
More Information
Set Membership: |
|

APL1
Genomic Map