Assay Details
| Gene Symbol: APL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121927.3 | NP_197423.1 | 2 - 3 | 395 | 119 | |
| GenBank mRNA | AF370503.1 | - | 2 - 3 | 394 | 119 | |
| BT008884.1 | - | 2 - 3 | 339 | 119 | ||
| U72290.1 | - | 2 - 3 | 352 | 119 | ||
| X73367.2 | - | 352 | 119 | |||
More Information
Set Membership: |
|

APL1
Genomic Map