Assay Details
| Gene Symbol: APL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121927.3 | NP_197423.1 | 9 - 10 | 1079 | 144 | |
| GenBank mRNA | AF370503.1 | - | 9 - 10 | 1078 | 144 | |
| BT008884.1 | - | 9 - 10 | 1023 | 144 | ||
| U72290.1 | - | 9 - 10 | 1036 | 144 | ||
| X73367.2 | - | 1036 | 144 | |||
More Information
Set Membership: |
|

APL1
Genomic Map