Assay Details
| Gene Symbol: COQ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336307.1 | NP_001324991.1 | 7 - 8 | 886 | 65 | |
| NM_128644.4 | NP_180649.1 | 6 - 7 | 736 | 65 | ||
| GenBank mRNA | AY045828.1 | - | 6 - 7 | 727 | 65 | |
| AY091366.1 | - | 6 - 7 | 619 | 65 | ||
More Information
Set Membership: |
|

COQ3
Genomic Map