Assay Details
| Gene Symbol: TCP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128662.2 | NP_565712.1 | 1 - 2 | 1639 | 80 | |
| GenBank mRNA | AY059945.1 | - | 1 - 2 | 1639 | 80 | |
| BT001194.1 | - | 1 - 2 | 1099 | 80 | ||
More Information
Set Membership: |
|

TCP10
Genomic Map