Assay Details
| Gene Symbol: CCT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336383.1 | NP_001323485.1 | 5 - 6 | 1260 | 71 | |
| NM_128785.3 | NP_180785.1 | 6 - 7 | 858 | 71 | ||
| GenBank mRNA | AY136393.1 | - | 6 - 7 | 791 | 71 | |
| BT000207.1 | - | 6 - 7 | 670 | 71 | ||
More Information
Set Membership: |
|

CCT1
Genomic Map