Assay Details
| Gene Symbol: NUDT22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336477.1 | NP_001325145.1 | 3 - 4 | 733 | 77 | |
| NM_001336478.1 | NP_001325146.1 | 3 - 4 | 803 | 77 | ||
| NM_001336479.1 | NP_001325147.1 | 3 - 4 | 713 | 77 | ||
| NM_128949.3 | NP_565776.1 | 3 - 4 | 646 | 77 | ||
| GenBank mRNA | AF360290.1 | - | 3 - 4 | 718 | 77 | |
| AK175540.1 | - | 3 - 4 | 644 | 77 | ||
| AK229595.1 | - | 3 - 4 | 680 | 77 | ||
| AY051046.1 | - | 3 - 4 | 585 | 77 | ||
More Information
Set Membership: |
|

NUDT22
Genomic Map