Assay Details
| Gene Symbol: ZML2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104038.4 | NP_564593.1 | 3 - 4 | 862 | 75 | |
| NM_202273.3 | NP_974002.1 | 3 - 4 | 624 | 75 | ||
| GenBank mRNA | AB119061.1 | - | 3 - 4 | 536 | 75 | |
| AY150395.1 | - | 2 - 3 | 395 | 75 | ||
More Information
Set Membership: |
|

ZML2
Genomic Map