Assay Details
| Gene Symbol: BGLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340171.1 | NP_001327167.1 | 11 - 12 | 1311 | 151 | |
| NM_116139.3 | NP_191833.2 | 11 - 12 | 1311 | 151 | ||
More Information
Set Membership: |
|

BGLU7
Genomic Map