Assay Details
| Gene Symbol: NPL41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116165.4 | NP_191859.1 | 1 - 2 | 1228 | 113 | |
| GenBank mRNA | AY035064.1 | - | 1 - 2 | 1100 | 113 | |
| AY056362.1 | - | 1 - 2 | 984 | 113 | ||
More Information
Set Membership: |
|

NPL41
Genomic Map