Assay Details
| Gene Symbol: LIF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036489.3 | NP_001031566.1 | 2 - 3 | 837 | 72 | |
| NM_001203718.2 | NP_001190647.2 | 2 - 3 | 653 | 72 | ||
| NM_001203719.2 | NP_001190648.1 | 2 - 3 | 836 | 72 | ||
| NM_001340269.1 | NP_001328329.1 | 2 - 3 | 822 | 72 | ||
| NM_001340270.1 | NP_001328330.1 | 1 - 2 | 460 | 72 | ||
| NM_116309.4 | NP_567192.1 | 2 - 3 | 658 | 72 | ||
| GenBank mRNA | AF367357.1 | - | 2 - 3 | 575 | 72 | |
| AK227578.1 | - | 2 - 3 | 764 | 72 | ||
| AY113171.1 | - | 1 - 2 | 430 | 72 | ||
More Information
Set Membership: |
|

LIF2
Genomic Map