Assay Details
| Gene Symbol: LIF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036489.3 | NP_001031566.1 | 5 - 6 | 1275 | 118 | |
| NM_001203718.2 | NP_001190647.2 | 5 - 6 | 1067 | 118 | ||
| NM_001203719.2 | NP_001190648.1 | 5 - 6 | 1274 | 118 | ||
| NM_001340269.1 | NP_001328329.1 | 5 - 6 | 1236 | 118 | ||
| NM_001340270.1 | NP_001328330.1 | 4 - 5 | 898 | 118 | ||
| NM_116309.4 | NP_567192.1 | 5 - 6 | 1096 | 118 | ||
| GenBank mRNA | AF367357.1 | - | 5 - 6 | 1013 | 118 | |
| AK227578.1 | - | 5 - 6 | 1202 | 118 | ||
| AY113171.1 | - | 4 - 5 | 868 | 118 | ||
More Information
Set Membership: |
|

LIF2
Genomic Map