Assay Details
| Gene Symbol: AT4G02030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203725.1 | NP_001190654.1 | 18 - 19 | 1911 | 133 | |
| NM_116434.4 | NP_192112.2 | 16 - 17 | 1930 | 133 | ||
| GenBank mRNA | AK228828.1 | - | 16 - 17 | 1836 | 133 | |
More Information
Set Membership: |
|

AT4G02030
Genomic Map