Assay Details
| Gene Symbol: RSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340369.1 | NP_001329795.1 | 3 - 4 | 709 | 104 | |
| NM_116459.4 | NP_567226.1 | 3 - 4 | 726 | 104 | ||
| NM_178956.4 | NP_849287.2 | 3 - 4 | 717 | 104 | ||
| NM_202772.2 | NP_974501.1 | 3 - 4 | 717 | 104 | ||
| GenBank mRNA | AF225702.1 | - | 3 - 4 | 570 | 104 | |
More Information
Set Membership: |
|

RSH1
Genomic Map