Assay Details
| Gene Symbol: RSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340369.1 | NP_001329795.1 | 9 - 10 | 1257 | 92 | |
| NM_116459.4 | NP_567226.1 | 9 - 10 | 1274 | 92 | ||
| NM_178956.4 | NP_849287.2 | 9 - 10 | 1265 | 92 | ||
| NM_202772.2 | NP_974501.1 | 9 - 10 | 1265 | 92 | ||
| GenBank mRNA | AF225702.1 | - | 9 - 10 | 1118 | 92 | |
More Information
Set Membership: |
|

RSH1
Genomic Map