Assay Details
| Gene Symbol: EMB3129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116513.5 | NP_192188.2 | 6 - 7 | 841 | 101 | |
| GenBank mRNA | AY150415.1 | - | 6 - 7 | 752 | 101 | |
More Information
Set Membership: |
|

EMB3129
Genomic Map