Assay Details
| Gene Symbol: EMB2369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116673.7 | NP_192344.1 | 3 - 4 | 625 | 66 | |
| GenBank mRNA | AY079159.1 | - | 3 - 4 | 625 | 66 | |
| BT002249.1 | - | 3 - 4 | 586 | 66 | ||
More Information
Set Membership: |
|

EMB2369
Genomic Map