Assay Details
| Gene Symbol: DDB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125468.1 | NP_001118940.1 | 10 - 11 | 1075 | 64 | |
| NM_116781.3 | NP_192451.1 | 11 - 12 | 1146 | 64 | ||
| GenBank mRNA | BT001905.1 | - | 11 - 12 | 972 | 64 | |
More Information
Set Membership: |
|

DDB1A
Genomic Map