Assay Details
| Gene Symbol: AT1G53210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104200.3 | NP_564623.2 | 4 - 5 | 1252 | 75 | |
| GenBank mRNA | AY099550.1 | - | 4 - 5 | 1070 | 75 | |
More Information
Set Membership: |
|

AT1G53210
Genomic Map