Assay Details
| Gene Symbol: TTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104208.3 | NP_175737.1 | 5 - 6 | 2159 | 95 | |
| GenBank mRNA | BT006444.1 | - | 5 - 6 | 1838 | 95 | |
More Information
Set Membership: |
|

TTL1
Genomic Map