Assay Details
| Gene Symbol: FAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104230.2 | NP_175757.1 | 1 - 2 | 362 | 77 | |
| GenBank mRNA | BT025725.1 | - | 1 - 2 | 229 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAP3| Gene Symbol: FAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104230.2 | NP_175757.1 | 1 - 2 | 362 | 77 | |
| GenBank mRNA | BT025725.1 | - | 1 - 2 | 229 | 77 | |
Set Membership: |
|