Assay Details
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333687.1 | NP_001320298.1 | 1 - 2 | 572 | 108 | |
| NM_104382.5 | NP_175905.2 | 1 - 2 | 554 | 108 | ||
| GenBank mRNA | AK227122.1 | - | 1 - 2 | 555 | 108 | |
| AY093241.1 | - | 1 - 2 | 511 | 108 | ||
| BT006281.1 | - | 1 - 2 | 474 | 108 | ||
More Information
Set Membership: |
|

MED9
Genomic Map