Assay Details
| Gene Symbol: MED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_104382.5 | NP_175905.2 | 3 - 4 | 699 | 106 | |
| GenBank mRNA | AK227122.1 | - | 3 - 4 | 700 | 106 | |
| AY093241.1 | - | 3 - 4 | 656 | 106 | ||
| BT006281.1 | - | 3 - 4 | 619 | 106 | ||
More Information
Set Membership: |
|

MED9
Genomic Map