Assay Details
| Gene Symbol: HUB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198304.1 | NP_001185233.1 | 2 - 3 | 467 | 96 | |
| NM_001333696.1 | NP_001321104.1 | 2 - 3 | 544 | 96 | ||
| NM_104398.4 | NP_564680.4 | 2 - 3 | 346 | 96 | ||
More Information
Set Membership: |
|

HUB2
Genomic Map